This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01158519_g1
See other APBB3 GE Assays ›
Gene Symbol:
Apbb3
Gene Name:
amyloid beta (A4) precursor protein-binding, family B, member 3
Gene Aliases:
Fe65l2, Rirl2, TR2S
Chromosome Location:
Chr.18: 36671154 - 36679366 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01158519_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Apbb3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_146085.1 NP_666197.1 9 - 10 1158 67
GenBank mRNA AK141878.1 - 9 - 10 1060 67
AK171634.1 - 8 - 9 1351 67
AK210931.1 - 2 - 3 133 67
BC024136.1 - 8 - 9 1156 67
BC024809.1 - 9 - 10 1158 5357681 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800001748.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Apbb3

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
negative regulation of apoptotic process
beta-amyloid binding
transcription factor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800008808.mm.1 NM_146085.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800008808.mm.1 NM_146085.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800008808.mm.1 TC1800001748.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7kcm7:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0