This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01158970_m1
See other BMX GE Assays ›
Gene Symbol:
Bmx
Gene Name:
BMX non-receptor tyrosine kinase
Gene Aliases:
Etk, Etk/Bmx, Tyro8
Chromosome Location:
Chr.X: 164192842 - 164258193 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01158970_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bmx
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009759.4 NP_033889.2 12 - 13 1230 112
GenBank mRNA AF012104.1 - 11 - 12 1083 112
AK036707.1 - 11 - 12 1089 112
AK040936.1 - 12 - 13 1231 112
BC138104.1 - 12 - 13 1107 8861123 112
BC138105.1 - 12 - 13 1107 8861124 112
U88091.1 - 12 - 13 1216 112

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00003331.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Bmx

Gene Ontology Categories:

Function(s) Process(es)

adaptive immune response
protein phosphorylation
apoptotic process
cell adhesion
signal transduction
transmembrane receptor protein tyrosine kinase signaling pathway
phosphorylation
peptidyl-tyrosine phosphorylation
cell differentiation
intracellular signal transduction
peptidyl-tyrosine autophosphorylation
regulation of cell proliferation
innate immune response
protein autophosphorylation
nucleotide binding
protein kinase activity
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00020757.mm.1 NM_009759.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00020757.mm.1 NM_009759.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00020757.mm.1 TC0X00003331.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0