This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01159012_m1
See other ACE2 GE Assays ›
Gene Symbol:
Ace2
Gene Name:
angiotensin I converting enzyme (peptidyl-dipeptidase A) 2
Gene Aliases:
2010305L05Rik
Chromosome Location:
Chr.X: 164139342 - 164188418 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01159012_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ace2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001130513.1 NP_001123985.1 7 - 8 1132 114
NM_027286.4 NP_081562.2 6 - 7 984 114
GenBank mRNA AB053181.1 - 6 - 7 988 114
AB053182.1 - 6 - 7 988 114
AK135357.1 - 1 - 2 166 114
AK141604.1 - 6 - 7 833 114
BC026801.1 - 6 - 7 977 4236529 114
EF408739.1 - 7 - 8 1093 114
EF408740.1 - 7 - 8 1093 114
GQ262785.1 - 6 - 7 802 114

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00001663.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ace2

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

angiotensin-mediated drinking behavior
regulation of systemic arterial blood pressure by renin-angiotensin
proteolysis
receptor biosynthetic process
viral entry into host cell
receptor-mediated virion attachment to host cell
negative regulation of smooth muscle cell proliferation
positive regulation of cardiac muscle contraction
negative regulation of ERK1 and ERK2 cascade
positive regulation of gap junction assembly
regulation of cardiac conduction
virus receptor activity
glycoprotein binding
endopeptidase activity
carboxypeptidase activity
peptidase activity
metallopeptidase activity
peptidyl-dipeptidase activity
hydrolase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0