This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01159651_m1
See other FAM173B GE Assays ›
Gene Symbol:
Fam173b
Gene Name:
family with sequence similarity 173, member B
Gene Aliases:
A930016P21Rik, AA987072
Chromosome Location:
Chr.15: 31601941 - 31617537 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01159651_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fam173b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026546.3 NP_080822.1 1 - 2 37 52
GenBank mRNA AK020868.1 - 1 - 2 37 52

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK020868.1, NM_026546.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fam173b

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500001543.mm.1 NM_026546.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500001543.mm.1 NM_026546.3 Refseq
Affymetrix Mouse Clariom S Array PSR1500001543.mm.1 TC1500000208.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline