This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01159654_g1
See other CCT5 GE Assays ›
Gene Symbol:
Cct5
Gene Name:
chaperonin containing Tcp1, subunit 5 (epsilon)
Gene Aliases:
Ccte, TCPE, mKIAA0098
Chromosome Location:
Chr.15: 31590813 - 31601804 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01159654_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cct5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007637.2 NP_031663.1 6 - 7 962 84
GenBank mRNA AK077753.1 - 6 - 7 920 84
AK088184.1 - 6 - 7 896 84
AK129056.1 - 6 - 7 868 84
AK133857.1 - 6 - 7 920 84
AK136493.1 - 6 - 7 914 84
AK145445.1 - 6 - 7 931 84
AK146405.1 - 6 - 7 934 84
AK146425.1 - 6 - 7 914 84
AK150390.1 - 6 - 7 881 84
AK153911.1 - 6 - 7 938 84
AK167895.1 - 6 - 7 903 84
BC092233.1 - 6 - 7 932 84
BC094427.1 - 6 - 7 909 30537966 84
Z31555.1 - 6 - 7 962 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500001326.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cct5

Panther Classification:

Molecular Function -

chaperonin

Gene Ontology Categories:

Function(s) Process(es)

protein folding
binding of sperm to zona pellucida
toxin transport
nucleotide binding
protein binding
ATP binding
G-protein beta-subunit binding
beta-tubulin binding
unfolded protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500010294.mm.1 NM_007637.3 Refseq
Affymetrix Mouse Clariom S Array PSR1500010294.mm.1 TC1500001326.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500010295.mm.1 NM_007637.3 Refseq
Affymetrix Mouse Clariom S Array PSR1500010295.mm.1 TC1500001326.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline