This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01159800_s1
See other GPR17 GE Assays ›
Gene Symbol:
Gpr17
Gene Name:
G protein-coupled receptor 17
Gene Aliases:
AI853548
Chromosome Location:
Chr.18: 31942999 - 31949636 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01159800_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr17
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001025381.2 NP_001020552.1 2 2284 67
GenBank mRNA BC070439.1 - 2 2284 30533246 67

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpr17

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
G-protein coupled receptor signaling pathway
signal transducer activity
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800008128.mm.1 NM_001025381.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800008128.mm.1 NM_001025381.2 Refseq
Affymetrix Mouse Clariom S Array PSR1800008128.mm.1 TC1800001119.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800008129.mm.1 NM_001025381.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800008129.mm.1 NM_001025381.2 Refseq
Affymetrix Mouse Clariom S Array PSR1800008129.mm.1 TC1800001119.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rzthw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0