Assay Details
| Gene Symbol: Tmem194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113211.1 | NP_001106682.1 | 4 - 5 | 566 | 92 | |
| NM_173732.3 | NP_776093.1 | 4 - 5 | 566 | 92 | ||
| GenBank mRNA | AK090031.1 | - | 4 - 5 | 545 | 92 | |
| AK129110.1 | - | 4 - 5 | 563 | 92 | ||
| AK132974.1 | - | 1035 | 92 | |||
| BC030440.1 | - | 4 - 5 | 577 | 5373527 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem194
Genomic Map