This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01160668_g1
See other STAT2 GE Assays ›
Gene Symbol:
Stat2
Gene Name:
signal transducer and activator of transcription 2
Gene Aliases:
1600010G07Rik, AW496480
Chromosome Location:
Chr.10: 128270559 - 128292852 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01160668_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stat2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019963.1 NP_064347.1 5 - 6 627 100
GenBank mRNA AF088862.1 - 5 - 6 497 100
AF187231.1 - 5 - 6 621 100
AF206162.1 - 5 - 6 627 100
AK149838.1 - 5 - 6 624 100
AK149924.1 - 5 - 6 640 100
AK150070.1 - 5 - 6 639 100
AK153405.1 - 3 - 4 265 100

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Stat2

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of defense response to virus by host
transcription, DNA-templated
regulation of transcription, DNA-templated
signal transduction
mitophagy in response to mitochondrial depolarization
DNA binding
transcription factor activity, sequence-specific DNA binding
signal transducer activity
protein binding
identical protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline