This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01160728_g1
See other GSTK1 GE Assays ›
Gene Symbol:
Gstk1
Gene Name:
glutathione S-transferase kappa 1
Gene Aliases:
0610025I19Rik, AW260476, DsbA-L
Chromosome Location:
Chr.6: 42245935 - 42250441 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01160728_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gstk1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029555.2 NP_083831.1 3 - 4 366 72
GenBank mRNA AK002661.1 - 3 - 4 364 72
AY279096.1 - 3 - 4 279 72
BC038151.1 - 3 - 4 293 72
BC051507.1 - 3 - 4 330 72
BC058163.1 - 3 - 4 299 5100572 72

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gstk1

Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

glutathione metabolic process
oxidation-reduction process
glutathione transferase activity
glutathione peroxidase activity
receptor binding
protein disulfide oxidoreductase activity
transferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0