This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01160731_g1
See other GSTK1 GE Assays ›
Gene Symbol:
Gstk1
Gene Name:
glutathione S-transferase kappa 1
Gene Aliases:
0610025I19Rik, AW260476, DsbA-L
Chromosome Location:
Chr.6: 42245935 - 42250441 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01160731_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gstk1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029555.2 NP_083831.1 7 - 8 721 80
GenBank mRNA AK002661.1 - 7 - 8 719 80
AY279096.1 - 7 - 8 634 80
BC038151.1 - 7 - 8 648 80
BC051507.1 - 7 - 8 762 80
BC058163.1 - 7 - 8 654 5100572 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600000454.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gstk1

Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

glutathione metabolic process
oxidation-reduction process
glutathione transferase activity
glutathione peroxidase activity
receptor binding
protein disulfide oxidoreductase activity
transferase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2051313 BC038151.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2051313 BC051507.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2051313 NM_029555.2 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 379226 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 837360 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 517679 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160744 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2051313 BC038151.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2051313 BC051507.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2051313 NM_029555.2 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P459240 AK002661.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24397081 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24397078 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24397072 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24397056 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24397070 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24397048 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24397077 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24397063 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24397058 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24397075 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24397064 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24397067 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24397080 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24397060 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24397066 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24397061 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24397082 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24397052 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24397076 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24397050 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24397051 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24397069 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P160744 - -
Whole Mouse Genome G4122A A_51_P459240 AK002661.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135901 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P160744 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05563390 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05095382 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05563398 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05563394 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05563403 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397077 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397041 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397052 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397038 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397097 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397148 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397084 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397141 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397046 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397161 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397152 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397134 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397113 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397028 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397107 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397066 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397102 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397032 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397145 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397026 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397137 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397076 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397037 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397127 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397095 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397099 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397105 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397083 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397088 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397142 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397050 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397067 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397082 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397078 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397055 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397072 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397144 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397043 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397092 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397070 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397091 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397112 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397048 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397146 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397080 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397085 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397034 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397063 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397033 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397029 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397051 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397109 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397159 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397061 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397058 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24397150 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24397097 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24397041 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24397083 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24397075 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05563398 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24397067 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24397109 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24397030 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24397080 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24397037 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05839911 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24397105 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05724084 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24397058 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05843737 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05563403 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05702191 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05563402 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24397043 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05280586 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05293535 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24397028 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24397107 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24397077 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24397034 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24397082 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24397045 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05798594 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24397092 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24397076 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24397050 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05798592 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24397048 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24397091 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24397099 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05563389 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05563394 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24397072 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05140692 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24397052 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24397095 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24397051 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24397038 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05846972 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24397085 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10537714 - -
Affymetrix Mouse Gene 1.0 ST Array 10537718 - -
Affymetrix Mouse Gene 1.0 ST Array 10537715 - -
Affymetrix Mouse Gene 1.0 ST Array 10537716 - -
Affymetrix Mouse Gene 1.0 ST Array 10537720 - -
Affymetrix Mouse Gene 1.0 ST Array 10537717 - -
Affymetrix Mouse Gene 1.0 ST Array 10537713 - -
Affymetrix Mouse Gene 1.0 ST Array 10537719 - -
Affymetrix Mouse Gene 1.0 ST Array 10537721 - -
Affymetrix HT MG-430 PM Array Plate 1452823_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10537712 - -
Mouse Genome 430 2.0 Array 1452823_PM_at - -
Mouse Genome 430 2.0 Array 1452823_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 76263_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 76263_PM_at - -
Affymetrix Mouse Clariom S Array PSR0600003610.mm.1 TC0600000454.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600003610.mm.1 NM_029555.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600003610.mm.1 NM_029555.2 Refseq
Mouse Expression Set 430 1452823_PM_at - -
Mouse Expression Set 430 1452823_at - -
Murine Genome U74v2 Set 96670_at - -
Murine Genome U74v2 Set 96670_PM_at - -
Mouse Genome 430A 2.0 Array 1452823_at - -
Mouse Genome 430A 2.0 Array 1452823_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1452823_at - -
Affymetrix HT Mouse Genome 430A Array 1452823_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 402550 - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 100730739 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2776008 - -
mouse-6 v1.1 (Illumina) scl076263.8_129-S - -
Illumina Sentrix mouse-6.v1 bead array 100730739 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 100730739 - -
MouseRef-8 v2 Expression BeadChip ILMN_2792924 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2792924 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2776008 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0