This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01161340_g1
See other AGER GE Assays ›
Gene Symbol:
Ager
Gene Name:
advanced glycosylation end product-specific receptor
Gene Aliases:
RAGE
Chromosome Location:
Chr.17: 34597410 - 34600937 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01161340_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ager
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001271422.1 NP_001258351.1 8 - 9 1001 95
NM_001271423.1 NP_001258352.1 8 - 9 974 95
NM_001271424.1 NP_001258353.1 7 - 8 1096 95
NR_073176.1 7 - 8 1196 95
GenBank mRNA AK004802.1 - 8 - 9 991 95
EU570243.1 - 8 - 9 957 95
EU570247.1 - 7 - 8 1152 95
EU906863.1 - 8 - 9 930 95
GU164727.1 - 7 - 8 1071 95
GU164729.1 - 7 - 8 812 95
GU164733.1 - 3 - 4 326 95

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Inventoried Probe spans exons

Panther Classification:

Molecular Function -

cell adhesion molecule immunoglobulin superfamily cell adhesion molecule

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
negative regulation of protein phosphorylation
positive regulation of protein phosphorylation
negative regulation of endothelial cell proliferation
inflammatory response
negative regulation of cell adhesion
JAK-STAT cascade
positive regulation of autophagy
negative regulation of endothelial cell migration
positive regulation of gene expression
positive regulation of epithelial to mesenchymal transition
positive regulation of fibroblast migration
positive regulation of smooth muscle cell migration
positive regulation of cell migration
neuron projection development
negative regulation of collagen biosynthetic process
positive regulation of apoptotic process
positive regulation of JUN kinase activity
positive regulation of neuron apoptotic process
positive regulation of fibroblast proliferation
positive regulation of smooth muscle cell proliferation
regulation of inflammatory response
positive regulation of inflammatory response
induction of positive chemotaxis
positive regulation of NF-kappaB transcription factor activity
regulation of DNA binding
calcium ion homeostasis
positive regulation of phagocytosis, engulfment
protein localization to membrane
positive regulation of potassium ion transmembrane transporter activity
positive regulation of neuron death
positive regulation of endothelial cell apoptotic process
positive regulation of reactive oxygen species metabolic process
positive regulation of type B pancreatic cell apoptotic process
protein binding
identical protein binding
S100 protein binding
advanced glycation end-product receptor activity
high mobility group box 1 binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0