This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01162214_m1
See other MON2 GE Assays ›
Gene Symbol:
Mon2
Gene Name:
MON2 homolog (yeast)
Gene Aliases:
2610528O22Rik, AW495628, Sf21, mKIAA1040
Chromosome Location:
Chr.10: 122992060 - 123076505 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01162214_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mon2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163024.1 NP_001156496.1 25 - 26 3746 105
NM_001163025.1 NP_001156497.1 24 - 25 3746 105
NM_153395.2 NP_700444.2 24 - 25 3743 105
GenBank mRNA AK011826.1 - 3 - 4 534 105
AK012182.2 - 4 - 5 683 105
AK122425.1 - 25 - 26 3746 105
AK162859.1 - 1 - 2 237 105
BC048406.1 - 12 - 13 1522 105
BC150943.1 - 24 - 25 3267 9087854 105
CV563363.1 - 3 - 4 606 105

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK011826.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mon2

Panther Classification:

Molecular Function -

guanyl-nucleotide exchange factor

Gene Ontology Categories:

Function(s) Process(es)

transport
protein transport
vesicle-mediated transport
positive regulation of GTPase activity
ARF guanyl-nucleotide exchange factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kjp6x:80/100.66.78.163:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0