This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01162536_m1
See other LRRC51 GE Assays ›
Gene Symbol:
Lrrc51
Gene Name:
leucine rich repeat containing 51
Gene Aliases:
1700008D07Rik, Lrtomt
Chromosome Location:
Chr.7: 101912989 - 101933917 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01162536_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lrrc51
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027053.1 NP_081329.1 1 - 2 89 77
GenBank mRNA AK005758.1 - 1 - 2 40 77

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK005758.1, NM_027053.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700032496.mm.1 NM_001162973.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700032496.mm.1 NM_001162973.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700032496.mm.1 NM_027053.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700032496.mm.1 NM_027053.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700032496.mm.1 TC0700003848.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700032493.mm.1 NM_001162973.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700032493.mm.1 NM_001162973.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700032493.mm.1 NM_027053.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700032493.mm.1 NM_027053.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700032493.mm.1 TC0700003848.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700032491.mm.1 NM_027053.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700032491.mm.1 NM_027053.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700032491.mm.1 TC0700003848.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline