This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01162547_g1
See other SLCO2B1 GE Assays ›
Gene Symbol:
Slco2b1
Gene Name:
solute carrier organic anion transporter family, member 2b1
Gene Aliases:
AI060904, AI852653, OATP-B, Slc21a9
Chromosome Location:
Chr.7: 99657804 - 99711340 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01162547_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slco2b1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001252530.1 NP_001239459.1 12 - 13 2141 107
NM_001252531.1 NP_001239460.1 12 - 13 2084 107
NM_175316.3 NP_780525.1 12 - 13 1887 107
GenBank mRNA AK033539.1 - 4 - 5 513 107
AK048120.1 - 12 - 13 1888 107
AK087682.1 - 12 - 13 2139 107
AK132394.1 - 12 - 13 2142 107
AK163542.1 - 12 - 13 2137 107
AK173049.1 - 10 - 11 1673 107
BC019209.1 - 12 - 13 1870 107
BC037089.1 - 12 - 13 1859 107
BC049251.1 - 12 - 13 1850 107
BC096485.1 - 12 - 13 1896 5101188 107

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700003804.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slco2b1

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
organic anion transport
bile acid and bile salt transport
sodium-independent icosanoid transport
anion transmembrane transport
organic acid transmembrane transport
transporter activity
organic anion transmembrane transporter activity
bile acid transmembrane transporter activity
sodium-independent organic anion transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700032012.mm.1 NM_001252530.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700032012.mm.1 NM_001252530.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700032012.mm.1 NM_001252531.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700032012.mm.1 NM_001252531.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700032012.mm.1 NM_175316.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700032012.mm.1 NM_175316.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700032012.mm.1 TC0700003804.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline