This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01164265_m1
See other HELB GE Assays ›
Gene Symbol:
Helb
Gene Name:
helicase (DNA) B
Gene Aliases:
AI447783, D10Ertd664e
Chromosome Location:
Chr.10: 120083608 - 120112965 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01164265_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Helb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_080446.2 NP_536694.2 2 - 3 674 69
GenBank mRNA AB048542.1 - 2 - 3 599 69
AK131122.1 - 2 - 3 696 69
BC068140.1 - 2 - 3 602 6843107 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000003018.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Helb

Panther Classification:

Molecular Function -

RNA helicase

Gene Ontology Categories:

Function(s) Process(es)

DNA replication
DNA-dependent DNA replication
DNA replication, synthesis of RNA primer
DNA duplex unwinding
nucleotide binding
ATP-dependent DNA helicase activity
helicase activity
ATP binding
hydrolase activity
single-stranded DNA-dependent ATP-dependent DNA helicase activity
ATP-dependent 5'-3' DNA helicase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000022550.mm.1 NM_080446.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000022550.mm.1 NM_080446.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000022550.mm.1 TC1000003018.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0