This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01164359_m1
See other ASXL3 GE Assays ›
Gene Symbol:
Asxl3
Gene Name:
additional sex combs like 3 (Drosophila)
Gene Aliases:
C230079D11Rik, D430002O22Rik, D930044O18Rik, Gm329, Gm945
Chromosome Location:
Chr.18: 22344077 - 22530227 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01164359_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Asxl3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001167777.1 NP_001161249.1 2 - 3 158 79
GenBank mRNA AK082659.1 - 2 - 3 158 79

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK082659.1, NM_001167777.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
biological_process
molecular_function
DNA binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800001227.mm.1 NM_001167777.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800001227.mm.1 NM_001167777.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800001227.mm.1 TC1800000171.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline