This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01164623_m1
See other C330008A17RIK GE Assays ›
Gene Symbol:
C330008A17Rik
Gene Name:
RIKEN cDNA C330008A17 gene
Gene Aliases:
-
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01164623_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C330008A17Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK021191.1 - 1 - 2 232 77
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY728756.1 - 1 - 2 227 Chr. 18 - 35913135 - 35916753 [+] on Build GRCm38 77

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream CXXC finger 5 Cxxc5 4930415K17Rik -51.4kb Sense Chr. 18 - 35828924 - 35861693 on NCBI Build GRCm38
Downstream pleckstrin and Sec7 domain containing 2 Psd2 6330404E20Rik,
AW125584,
EFA6C
47.1kb Sense Chr. 18 - 35963881 - 36014715 on NCBI Build GRCm38

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

MicroArray Probes:

Agilent ABI

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 3971 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118218 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13814388 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13780497 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13814392 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13916217 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620819 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620825 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620894 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620803 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620813 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620839 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620807 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620888 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620845 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620842 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620823 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620817 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620804 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620862 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620891 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620810 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13916242 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14067617 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13916219 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620890 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13643948 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620924 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620827 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13916246 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620815 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13916229 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620920 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13804559 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620831 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13761713 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620837 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620824 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620897 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620857 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13916223 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620847 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620900 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620858 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620874 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13756297 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14125532 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620870 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620855 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620873 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620893 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620859 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13966334 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620832 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620906 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620808 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620841 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620885 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620843 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13916214 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P14067603 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620853 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620849 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620876 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620910 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620911 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13860588 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13960832 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P31620865 - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 597529 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-ndfxr:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline