This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01164680_m1
See other MAPRE2 GE Assays ›
Gene Symbol:
Mapre2
Gene Name:
microtubule-associated protein, RP/EB family, member 2
Gene Aliases:
AI314113, C820009F03Rik, D18Abb1e, EB1, EB2, RP1
Chromosome Location:
Chr.18: 23752333 - 23893861 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01164680_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mapre2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001162941.1 NP_001156413.1 1 - 2 148 78
GenBank mRNA AK028823.1 - 1 - 2 149 78

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK028823.1, NM_001162941.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

non-motor microtubule binding protein

Gene Ontology Categories:

Function(s) Process(es)

cell cycle
mitotic nuclear division
cell division
protein binding
microtubule binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800001320.mm.1 NM_001162941.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800001320.mm.1 NM_001162941.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800001320.mm.1 TC1800000184.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800001321.mm.1 NM_001162941.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800001321.mm.1 NM_001162941.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800001321.mm.1 NM_001162942.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800001321.mm.1 NM_001162942.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800001321.mm.1 TC1800000184.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-7669c5b596-vc5lg:80/100.66.78.61:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0