This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01164681_m1
See other MAPRE2 GE Assays ›
Gene Symbol:
Mapre2
Gene Name:
microtubule-associated protein, RP/EB family, member 2
Gene Aliases:
AI314113, C820009F03Rik, D18Abb1e, EB1, EB2, RP1
Chromosome Location:
Chr.18: 23752333 - 23893861 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01164681_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mapre2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001162941.1 NP_001156413.1 2 - 3 200 66
NM_001162942.1 NP_001156414.1 2 - 3 143 66
GenBank mRNA AI325879.1 - 2 - 3 137 66
AK028823.1 - 2 - 3 201 66
AK168834.1 - 2 - 3 85 66

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK168834.1, NM_001162942.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

non-motor microtubule binding protein

Gene Ontology Categories:

Function(s) Process(es)

cell cycle
mitotic nuclear division
cell division
protein binding
microtubule binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800001317.mm.1 NM_001162942.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800001317.mm.1 NM_001162942.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800001317.mm.1 TC1800000184.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800001318.mm.1 NM_001162942.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800001318.mm.1 NM_001162942.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800001318.mm.1 TC1800000184.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-976xq:80/100.66.77.211:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline