This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01165353_m1
See other CDH12 GE Assays ›
Gene Symbol:
Cdh12
Gene Name:
cadherin 12
Gene Aliases:
Cdhb
Chromosome Location:
Chr.15: 20448879 - 21597005 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01165353_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdh12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001008420.2 NP_001008420.1 5 - 6 1023 122
GenBank mRNA BC086655.1 - 5 - 6 1023 30542783 122

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500000139.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cdh12

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
calcium ion binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500001146.mm.1 NM_001008420.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500001146.mm.1 NM_001008420.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500001146.mm.1 TC1500000139.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500001149.mm.1 NM_001008420.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500001149.mm.1 NM_001008420.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500001149.mm.1 TC1500000139.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500001144.mm.1 NM_001008420.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500001144.mm.1 NM_001008420.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500001144.mm.1 TC1500000139.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500001145.mm.1 NM_001008420.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500001145.mm.1 NM_001008420.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500001145.mm.1 TC1500000139.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500001147.mm.1 NM_001008420.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500001147.mm.1 NM_001008420.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500001147.mm.1 TC1500000139.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500001148.mm.1 NM_001008420.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500001148.mm.1 NM_001008420.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500001148.mm.1 TC1500000139.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline