This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01166981_g1
See other GPR85 GE Assays ›
Gene Symbol:
Gpr85
Gene Name:
G protein-coupled receptor 85
Gene Aliases:
2900026B03Rik
Chromosome Location:
Chr.6: 13834454 - 13839937 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01166981_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr85
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145066.4 NP_659503.1 2 - 3 829 122
GenBank mRNA BC065154.1 - 2 - 3 829 6418997 122

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC065154.1, NM_145066.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpr85

Panther Classification:

Molecular Function -

G-protein coupled receptor transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
G-protein coupled receptor signaling pathway
intracellular signal transduction
signal transducer activity
G-protein coupled receptor activity
receptor signaling protein activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600014791.mm.1 NM_145066.5 Refseq
Affymetrix Mouse Clariom S Array PSR0600014791.mm.1 TC0600001877.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600014792.mm.1 NM_145066.5 Refseq
Affymetrix Mouse Clariom S Array PSR0600014792.mm.1 TC0600001877.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline