This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01168088_g1
See other UBE2H GE Assays ›
Gene Symbol:
Ube2h
Gene Name:
ubiquitin-conjugating enzyme E2H
Gene Aliases:
1500009C23Rik, AI181839, AI326965, AI462483, AW227540, E2-20K, Gid3, N28148, Ubc8
Chromosome Location:
Chr.6: 30211289 - 30304539 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01168088_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ube2h
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001169577.1 NP_001163048.1 3 - 4 213 107
NM_009459.3 NP_033485.1 5 - 6 730 107
GenBank mRNA AK131688.1 - 5 - 6 708 107
BC008517.1 - 5 - 6 536 3600233 107
U19854.1 - 5 - 6 459 107

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ube2h

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
protein K48-linked ubiquitination
protein K11-linked ubiquitination
nucleotide binding
ubiquitin-protein transferase activity
ATP binding
ligase activity
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600015693.mm.1 TC0600001992.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600015693.mm.1 NM_001169576.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600015693.mm.1 NM_001169576.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600015693.mm.1 NM_009459.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600015693.mm.1 NM_009459.3 Refseq
Affymetrix Mouse Clariom S Array PSR0600015691.mm.1 TC0600001992.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600015691.mm.1 NM_001169577.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600015691.mm.1 NM_001169577.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600015695.mm.1 TC0600001992.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600015695.mm.1 NM_001169576.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600015695.mm.1 NM_001169576.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600015695.mm.1 NM_009459.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600015695.mm.1 NM_009459.3 Refseq
Affymetrix Mouse Clariom S Array PSR0600015690.mm.1 TC0600001992.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600015690.mm.1 NM_001169577.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600015690.mm.1 NM_001169577.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600015690.mm.1 NM_009459.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600015690.mm.1 NM_009459.3 Refseq
Affymetrix Mouse Clariom S Array PSR0600015689.mm.1 TC0600001992.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600015689.mm.1 NM_001169577.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600015689.mm.1 NM_001169577.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600015689.mm.1 NM_009459.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600015689.mm.1 NM_009459.3 Refseq
Affymetrix Mouse Clariom S Array PSR0600015692.mm.1 TC0600001992.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600015692.mm.1 NM_001169576.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600015692.mm.1 NM_001169576.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600015692.mm.1 NM_009459.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600015692.mm.1 NM_009459.3 Refseq
Affymetrix Mouse Clariom S Array PSR0600015688.mm.1 TC0600001992.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600015688.mm.1 NM_001169576.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600015688.mm.1 NM_001169576.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600015688.mm.1 NM_001169577.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600015688.mm.1 NM_001169577.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600015688.mm.1 NM_009459.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600015688.mm.1 NM_009459.3 Refseq
Affymetrix Mouse Clariom S Array PSR0600015694.mm.1 TC0600001992.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600015694.mm.1 NM_001169576.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600015694.mm.1 NM_001169576.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600015694.mm.1 NM_009459.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600015694.mm.1 NM_009459.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0