This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01168386_m1
See other MMP8 GE Assays ›
Gene Symbol:
Mmp8
Gene Name:
matrix metallopeptidase 8
Gene Aliases:
BB138268
Chromosome Location:
Chr.9: 7558429 - 7568486 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01168386_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mmp8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008611.4 NP_032637.3 4 - 5 724 88
GenBank mRNA AK089234.1 - 4 - 5 695 88
AK089336.1 - 4 - 5 699 88
AK089381.1 - 4 - 5 696 88
AK089516.1 - 4 - 5 685 88
AK137468.1 - 4 - 5 696 88
AK154937.1 - 4 - 5 695 88
AK155740.1 - 4 - 5 695 88
AK155832.1 - 4 - 5 695 88
AK155901.1 - 4 - 5 698 88
AK170177.1 - 4 - 5 695 88
AK170388.1 - 4 - 5 697 88
AK170760.1 - 4 - 5 695 88
AK171168.1 - 4 - 5 695 88
AK171613.1 - 4 - 5 697 88
BC042742.1 - 4 - 5 705 5032770 88
U96696.1 - 4 - 5 690 88
Y13342.1 - 4 - 5 622 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900000050.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mmp8

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
collagen catabolic process
metalloendopeptidase activity
serine-type endopeptidase activity
calcium ion binding
peptidase activity
metallopeptidase activity
zinc ion binding
hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900000405.mm.1 NM_008611.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900000405.mm.1 NM_008611.4 Refseq
Affymetrix Mouse Clariom S Array PSR0900000405.mm.1 TC0900000050.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nbrrd:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0