This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01168528_g1
See other HINFP GE Assays ›
Gene Symbol:
Hinfp
Gene Name:
histone H4 transcription factor
Gene Aliases:
AA589481, HiNF-P, Mizf
Chromosome Location:
Chr.9: 44292438 - 44305705 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01168528_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hinfp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172162.3 NP_751894.2 7 - 8 930 65
GenBank mRNA AK049348.1 - 7 - 8 892 65
AK049633.1 - 7 - 8 1091 65
BC029091.1 - 6 - 7 913 2651006 65
CT010239.1 - 6 - 7 865 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900002155.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hinfp

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

DNA damage checkpoint
G1/S transition of mitotic cell cycle
regulation of transcription involved in G1/S transition of mitotic cell cycle
negative regulation of transcription from RNA polymerase II promoter
in utero embryonic development
DNA repair
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
regulation of gene expression
positive regulation of gene expression
negative regulation of gene expression
establishment of protein localization
myoblast differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
nucleic acid binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
enzyme binding
histone binding
transcription regulatory region DNA binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900017889.mm.1 NM_172162.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900017889.mm.1 NM_172162.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900017889.mm.1 TC0900002155.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900017887.mm.1 NM_172162.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900017887.mm.1 NM_172162.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900017887.mm.1 TC0900002155.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900017888.mm.1 NM_172162.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900017888.mm.1 NM_172162.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900017888.mm.1 TC0900002155.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline