This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01168770_m1
See other PLA2G12B GE Assays ›
Gene Symbol:
Pla2g12b
Gene Name:
phospholipase A2, group XIIB
Gene Aliases:
2010002E04Rik, Fksg71, Pla2g13, hlb218
Chromosome Location:
Chr.10: 59403420 - 59421976 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01168770_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pla2g12b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_023530.2 NP_076019.2 3 - 4 553 59
GenBank mRNA AK008035.1 - 3 - 4 579 59
AY358033.1 - 3 - 4 582 59
BC021592.1 - 3 - 4 553 5133141 59

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000000534.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pla2g12b

Panther Classification:

Molecular Function -

phospholipase

Gene Ontology Categories:

Function(s) Process(es)

lipid catabolic process
cholesterol homeostasis
triglyceride homeostasis
molecular_function
phospholipase A2 activity
calcium ion binding
metal ion binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27168474 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P184385 NM_023530.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P184385 AF339738.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P184385 AK008035.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P184385 AY358033.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P184385 BC021592.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P184385 NM_023530.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P184385 AF339738.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P184385 AK008035.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P184385 AY358033.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P184385 BC021592.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 854556 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 128225 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 720346 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 433567 AK052791.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 33374 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 396732 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 84390 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 800076 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 542175 AK013690.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 542175 BC027647.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 542175 BC038883.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 542175 BC065420.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 542175 NM_024448.2 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 542175 XM_001478686.1 refseq_mrna_mouse_XM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 487075 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 170863 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 142376 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 153076 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P126133 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174148 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P184385 NM_023530.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P184385 AF339738.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P184385 AK008035.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P184385 AY358033.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P184385 BC021592.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P184385 NM_023530.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P184385 AF339738.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P184385 AK008035.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P184385 AY358033.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P184385 BC021592.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P184385 NM_023530.2 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P184385 AF339738.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P184385 AK008035.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P184385 AY358033.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P184385 BC021592.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P184385 NM_023530.2 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P184385 AF339738.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P184385 AK008035.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P184385 AY358033.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P184385 BC021592.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27168503 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27168508 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27168497 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27168506 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27168512 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27168514 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27168496 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27168501 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27168515 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27168504 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27168500 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27168498 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27168492 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27168494 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27168511 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07072641 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02134160 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P126133 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P174148 - -
Whole Mouse Genome G4122A A_51_P184385 NM_023530.2 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P184385 AF339738.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P184385 AK008035.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P184385 AY358033.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P184385 BC021592.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P184385 NM_023530.2 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P184385 AF339738.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P184385 AK008035.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P184385 AY358033.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P184385 BC021592.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121123 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P174148 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P126133 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08402829 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08402837 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08402836 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08402838 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08402830 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08402825 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27168474 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168584 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168562 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168501 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168613 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168535 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168537 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168533 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168636 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168572 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168603 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168639 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168605 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168558 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168504 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168607 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168635 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168628 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168497 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168641 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168487 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168567 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168626 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168570 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168545 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168648 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168588 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168486 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168522 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168582 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168555 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168470 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168593 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168541 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168542 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168622 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168538 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168653 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168612 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168590 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168569 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168644 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168526 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168508 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168496 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168547 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168571 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168610 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168520 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168634 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168544 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168616 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168620 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168551 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168565 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168617 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168645 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168529 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168511 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168665 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168608 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168473 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168643 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168621 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168583 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168548 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168647 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168506 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168503 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168602 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168627 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168580 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168629 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168518 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168658 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168512 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168498 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168624 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168560 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168661 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168527 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168663 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168524 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168614 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168500 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168532 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168514 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168651 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27168553 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27168491 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27168500 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27168524 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27168527 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27168514 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27168497 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27168520 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27168502 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27168508 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27168501 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27168535 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27168517 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08329933 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27168473 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27168518 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27168538 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27168522 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27168474 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08679327 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27168534 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08402838 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27168486 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27168511 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27168495 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27168504 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27168512 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27168506 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08234399 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27168496 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27168537 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27168541 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27168542 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08607896 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27168533 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27168529 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27168487 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08535392 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27168471 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10363376 - -
Affymetrix Mouse Gene 1.0 ST Array 10363373 - -
Affymetrix Mouse Gene 1.0 ST Array 10363375 - -
Affymetrix Mouse Gene 1.0 ST Array 10363374 - -
Affymetrix HT MG-430 PM Array Plate 1419614_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10363372 - -
Mouse Genome 430 2.0 Array 1419614_at - -
Mouse Genome 430 2.0 Array 1419614_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 69836_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 69836_PM_at - -
Affymetrix Mouse Clariom S Array PSR1000003566.mm.1 NM_023530.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000003566.mm.1 NM_023530.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000003566.mm.1 TC1000000534.mm.1 MTA TC
Mouse Expression Set 430 1419614_at - -
Mouse Expression Set 430 1419614_PM_at - -
Mouse Genome 430A 2.0 Array 1419614_at - -
Mouse Genome 430A 2.0 Array 1419614_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1419614_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1419614_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 698259 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 3290484 - -
Illumina mouse-6 v1.0 expression beadchip 3290484 - -
Illumina mouseRef-8 v1.0 expression beadchip 0003290484 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2699466 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2699466 - -
mouse-6 v1.1 (Illumina) scl38889.4.92_69-S - -
Illumina Sentrix mouse-6.v1 bead array 3290484 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl38889.4.92_69-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 3290484 - -
MouseRef-8 v2 Expression BeadChip ILMN_2699466 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2699466 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0