This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01169071_m1
See other SUPV3L1 GE Assays ›
Gene Symbol:
Supv3l1
Gene Name:
suppressor of var1, 3-like 1 (S. cerevisiae)
Gene Aliases:
6330443E10Rik
Chromosome Location:
Chr.10: 62429329 - 62451294 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01169071_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Supv3l1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_181423.2 NP_852088.1 10 - 11 1310 82
GenBank mRNA AJ968954.1 - 10 - 11 1337 82
AK031911.1 - 10 - 11 1317 82
AK143755.1 - 10 - 11 1335 82
BC049796.1 - 10 - 11 1310 6332552 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000002258.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Supv3l1

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

mitochondrial mRNA catabolic process
positive regulation of mitochondrial RNA catabolic process
mitochondrial RNA 3'-end processing
DNA recombination
RNA catabolic process
positive regulation of cell growth
DNA duplex unwinding
mitochondrial ncRNA surveillance
mitochondrial mRNA surveillance
negative regulation of apoptotic process
mitochondrion morphogenesis
chromatin maintenance
mitochondrial RNA surveillance
nucleotide binding
DNA binding
DNA helicase activity
double-stranded RNA binding
ATP-dependent RNA helicase activity
helicase activity
ATP binding
hydrolase activity
hydrolase activity, acting on acid anhydrides
3'-5' RNA helicase activity
protein homodimerization activity
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000016092.mm.1 NM_181423.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000016092.mm.1 NM_181423.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000016092.mm.1 TC1000002258.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline