This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01169091_g1
See other DNA2 GE Assays ›
Gene Symbol:
Dna2
Gene Name:
DNA replication helicase 2 homolog (yeast)
Gene Aliases:
Dna2l, E130315B21Rik
Chromosome Location:
Chr.10: 62946980 - 62974188 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01169091_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dna2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177372.3 NP_796346.2 11 - 12 1832 101
GenBank mRNA AK028381.1 - 11 - 12 1809 101
AK129051.1 - 11 - 12 1817 101
BC025182.1 - 6 - 7 983 101
BC030382.1 - 11 - 12 1773 101
BC115716.1 - 11 - 12 1786 40085888 101

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000000592.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dna2

Gene Ontology Categories:

Function(s) Process(es)

DNA replication checkpoint
DNA double-strand break processing
DNA replication
mitochondrial DNA replication
DNA repair
base-excision repair
cellular response to DNA damage stimulus
metabolic process
DNA duplex unwinding
DNA replication, Okazaki fragment processing
DNA replication, removal of RNA primer
mitochondrial DNA repair
positive regulation of DNA replication
nucleic acid phosphodiester bond hydrolysis
nucleotide binding
DNA binding
catalytic activity
helicase activity
nuclease activity
endonuclease activity
protein binding
ATP binding
hydrolase activity
site-specific endodeoxyribonuclease activity, specific for altered base
5'-flap endonuclease activity
phosphoric ester hydrolase activity
5'-3' DNA helicase activity
single-stranded DNA-dependent ATPase activity
T/G mismatch-specific endonuclease activity
retroviral 3' processing activity
metal ion binding
iron-sulfur cluster binding
4 iron, 4 sulfur cluster binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000003999.mm.1 NM_177372.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000003999.mm.1 NM_177372.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000003999.mm.1 TC1000000592.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0