Assay Details
| Gene Symbol: Midn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB036882.1 | - | 4 - 5 | 847 | 117 | |
| AK154535.1 | - | 3 - 4 | 791 | 117 | ||
| BC034719.1 | - | 4 - 5 | 817 | 3710931 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|

Midn
Genomic Map