This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01169252_g1
See other SPINT2 GE Assays ›
Gene Symbol:
Spint2
Gene Name:
serine protease inhibitor, Kunitz type 2
Gene Aliases:
AL024025, C76321, HAI-2
Chromosome Location:
Chr.7: 29256330 - 29282064 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01169252_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Spint2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001082548.1 NP_001076017.1 4 - 5 738 61
NM_011464.2 NP_035594.1 5 - 6 909 61
GenBank mRNA AK007792.1 - 4 - 5 408 61
BC003431.1 - 4 - 5 671 2646512 61
BC026419.1 - 4 - 5 610 4219482 61
BQ174562.1 - 2 - 3 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700002720.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Spint2

Panther Classification:

Molecular Function -

protease inhibitor

Gene Ontology Categories:

Function(s) Process(es)

neural tube closure
establishment or maintenance of cell polarity
negative regulation of peptidase activity
negative regulation of endopeptidase activity
negative regulation of cell-cell adhesion
epithelial cell morphogenesis involved in placental branching
basement membrane organization
negative regulation of cell motility
serine-type endopeptidase inhibitor activity
peptidase inhibitor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700024056.mm.1 NM_001082548.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700024056.mm.1 NM_001082548.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700024056.mm.1 NM_011464.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700024056.mm.1 NM_011464.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700024056.mm.1 TC0700002720.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline