This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01169292_g1
See other ME1 GE Assays ›
Gene Symbol:
Me1
Gene Name:
malic enzyme 1, NADP(+)-dependent, cytosolic
Gene Aliases:
D9Ertd267e, Mdh-1, Mod-1, Mod1
Chromosome Location:
Chr.9: 86581363 - 86695914 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01169292_g1
Size
Availability Made To Order
Catalog # 4351372
Price 417.00
Your Price
Online offer: 387.65
387.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Me1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001198933.1 NP_001185862.1 5 - 6 838 77
NM_008615.2 NP_032641.2 5 - 6 760 77
GenBank mRNA AK004980.1 - 5 - 6 626 77
AK077968.1 - 5 - 6 628 77
AK163278.1 - 5 - 6 838 77
AK163403.1 - 5 - 6 838 77
BC003287.1 - 5 - 6 617 3582779 77
BC011081.1 - 5 - 6 658 3978265 77
BC080660.1 - 5 - 6 616 4240732 77
BC094028.1 - 5 - 6 615 4974808 77
J02652.1 - 5 - 6 661 77
M26756.1 - 5 - 6 603 77

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Me1

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

malate metabolic process
response to hormone
protein tetramerization
oxidation-reduction process
regulation of NADP metabolic process
malic enzyme activity
malate dehydrogenase (decarboxylating) (NAD+) activity
malate dehydrogenase (decarboxylating) (NADP+) activity
protein binding
oxaloacetate decarboxylase activity
oxidoreductase activity
manganese ion binding
metal ion binding
NADP binding
NAD binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-7669c5b596-4vr89:80/100.66.79.40:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0