This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01169453_m1
See other GLB1L3 GE Assays ›
Gene Symbol:
Glb1l3
Gene Name:
galactosidase, beta 1 like 3
Gene Aliases:
4921509F24Rik
Chromosome Location:
Chr.9: 26817953 - 26862100 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01169453_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Glb1l3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001113323.1 NP_001106794.1 12 - 13 1395 66
GenBank mRNA AK014852.1 - 12 - 13 1228 66
BC132200.1 - 12 - 13 1188 40130477 66
BC132202.1 - 12 - 13 1188 40130479 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900001940.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

galactosidase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
metabolic process
hydrolase activity, hydrolyzing O-glycosyl compounds
beta-galactosidase activity
lytic transglycosylase activity
mannosidase activity
mannosyl-oligosaccharide mannosidase activity
galactosidase activity
glucosidase activity
trehalase activity
fucosidase activity
hexosaminidase activity
amylase activity
hydrolase activity
hydrolase activity, acting on glycosyl bonds
dextrin alpha-glucosidase activity
starch alpha-glucosidase activity
beta-glucanase activity
beta-6-sulfate-N-acetylglucosaminidase activity
glucan endo-1,4-beta-glucosidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900016284.mm.1 NM_001113323.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016284.mm.1 NM_001113323.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016284.mm.1 TC0900001940.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900016283.mm.1 NM_001113323.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016283.mm.1 NM_001113323.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016283.mm.1 TC0900001940.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900016282.mm.1 NM_001113323.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016282.mm.1 NM_001113323.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016282.mm.1 TC0900001940.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-f5rhb:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0