This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01169495_m1
See other ATP5D GE Assays ›
Gene Symbol:
Atp5d
Gene Name:
ATP synthase, H+ transporting, mitochondrial F1 complex, delta subunit
Gene Aliases:
0610008F14Rik, 1500000I11Rik, AA960090, AI876556, AU020773, C85518
Chromosome Location:
Chr.10: 80142280 - 80145818 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01169495_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atp5d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025313.2 NP_079589.2 1 - 2 292 64
GenBank mRNA AA409366.1 - 1 - 2 296 64
AK002330.1 - 1 - 2 242 64
AK011374.1 - 1 - 2 213 64
AK018011.1 - 1 - 2 238 64
AK076064.1 - 1 - 2 217 64
AK151151.1 - 1 - 2 227 64
AK155285.1 - 1 - 2 276 64
AK165932.1 - 1 - 2 241 64
BC008273.1 - 1 - 2 201 2811419 64
CT010177.1 - 1 - 2 141 64

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atp5d

Panther Classification:

Molecular Function -

ATP synthase

Gene Ontology Categories:

Function(s) Process(es)

ATP biosynthetic process
transport
ion transport
ATP synthesis coupled proton transport
proton transport
ATP metabolic process
hydrogen ion transmembrane transport
hydrogen ion transmembrane transporter activity
ATPase activity
protein complex binding
proton-transporting ATP synthase activity, rotational mechanism
proton-transporting ATPase activity, rotational mechanism

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-6dxlp:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0