This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01169584_m1
See other NAA60 GE Assays ›
Gene Symbol:
Naa60
Gene Name:
N(alpha)-acetyltransferase 60, NatF catalytic subunit
Gene Aliases:
1200013P24Rik, AI315146, Nat15
Chromosome Location:
Chr.16: 3872501 - 3904781 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01169584_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Naa60
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290689.1 NP_001277618.1 2 - 3 323 126
NM_029090.3 NP_083366.1 3 - 4 439 126
GenBank mRNA AK004750.1 - 3 - 4 332 126
AK088736.1 - 3 - 4 357 126
BC004837.1 - 3 - 4 375 2654387 126
BY255312.1 - 3 - 4 357 126

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001290689.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Naa60

Panther Classification:

Molecular Function -

acetyltransferase

Gene Ontology Categories:

Function(s) Process(es)

nucleosome assembly
chromosome segregation
cell proliferation
chromatin modification
N-terminal peptidyl-methionine acetylation
histone H4 acetylation
histone acetyltransferase activity
peptide alpha-N-acetyltransferase activity
N-acetyltransferase activity
H4 histone acetyltransferase activity
transferase activity
transferase activity, transferring acyl groups

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600000089.mm.1 NM_029090.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600000089.mm.1 NM_029090.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600000089.mm.1 TC1600002166.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600000092.mm.1 NM_029090.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600000092.mm.1 NM_029090.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600000092.mm.1 TC1600002166.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0