This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01169773_m1
See other ALCAM GE Assays ›
Gene Symbol:
Alcam
Gene Name:
activated leukocyte cell adhesion molecule
Gene Aliases:
AI853494, BEN, CD166, DM-GRASP, MuSC, SC1
Chromosome Location:
Chr.16: 52248996 - 52453255 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01169773_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Alcam
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009655.2 NP_033785.1 2 - 3 708 109
GenBank mRNA AB003026.1 - 2 - 3 370 109
AK030851.1 - 2 - 3 709 109
AK031391.1 - 1784 109
AY644766.1 - 2 - 3 365 109
BC027280.1 - 2 - 3 365 3501215 109
U95030.1 - 2 - 3 176 109

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600001722.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Alcam

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
axon guidance
motor neuron axon guidance
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0