This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01170332_m1
See other ANLN GE Assays ›
Gene Symbol:
Anln
Gene Name:
anillin, actin binding protein
Gene Aliases:
1110037A17Rik, 2900037I21Rik, C78101, Scraps
Chromosome Location:
Chr.9: 22331214 - 22389696 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01170332_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Anln
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028390.3 NP_082666.2 6 - 7 1488 70
GenBank mRNA AK013624.1 - 3 - 4 532 70
AK038403.1 - 3 - 4 563 70
AK045703.1 - 3 - 4 452 70
BC027141.1 - 6 - 7 1452 70
BC032164.1 - 6 - 7 1506 5346703 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900001908.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Anln

Panther Classification:

Molecular Function -

non-motor actin binding protein

Gene Ontology Categories:

Function(s) Process(es)

mitotic cytokinesis
hematopoietic progenitor cell differentiation
cell cycle
mitotic nuclear division
cell division
glomerular visceral epithelial cell migration
actin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900016086.mm.1 NM_028390.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016086.mm.1 NM_028390.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900016086.mm.1 TC0900001908.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900016087.mm.1 NM_028390.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016087.mm.1 NM_028390.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900016087.mm.1 TC0900001908.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-v82xx:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0