This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01170665_m1
See other CCT8 GE Assays ›
Gene Symbol:
Cct8
Gene Name:
chaperonin containing Tcp1, subunit 8 (theta)
Gene Aliases:
AI132397, Cctq, Tcpq
Chromosome Location:
Chr.16: 87483325 - 87495869 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01170665_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cct8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009840.3 NP_033970.3 13 - 14 1527 131
GenBank mRNA AK014005.1 - 6 - 7 593 131
AK075711.1 - 13 - 14 1508 131
AK075880.1 - 13 - 14 1516 131
AK145661.1 - 13 - 14 1511 131
AK145754.1 - 13 - 14 1510 131
AK145918.1 - 13 - 14 1529 131
AK147038.1 - 13 - 14 1530 131
AK159969.1 - 13 - 14 1518 131
AK168465.1 - 13 - 14 1501 131
AK172867.1 - 13 - 14 1498 131
BC009007.1 - 13 - 14 1485 3582639 131
Z37164.1 - 13 - 14 1531 131

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600001986.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cct8

Panther Classification:

Molecular Function -

chaperonin

Gene Ontology Categories:

Function(s) Process(es)

protein folding
binding of sperm to zona pellucida
pore complex assembly
toxin transport
nucleotide binding
protein binding
ATP binding
unfolded protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600015419.mm.1 NM_009840.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600015419.mm.1 NM_009840.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600015419.mm.1 TC1600001986.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline