This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01170668_m1
See other CCT8 GE Assays ›
Gene Symbol:
Cct8
Gene Name:
chaperonin containing Tcp1, subunit 8 (theta)
Gene Aliases:
AI132397, Cctq, Tcpq
Chromosome Location:
Chr.16: 87483325 - 87495869 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01170668_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cct8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009840.3 NP_033970.3 7 - 8 849 66
GenBank mRNA AK010882.1 - 7 - 8 850 66
AK075711.1 - 7 - 8 830 66
AK075880.1 - 7 - 8 838 66
AK145661.1 - 7 - 8 833 66
AK145754.1 - 7 - 8 832 66
AK145918.1 - 7 - 8 851 66
AK147038.1 - 7 - 8 852 66
AK159969.1 - 7 - 8 841 66
AK168465.1 - 7 - 8 822 66
AK172867.1 - 7 - 8 820 66
AK191019.1 - 3 - 4 239 66
AK211232.1 - 3 - 4 239 66
AK214517.1 - 3 - 4 255 66
AK219341.1 - 3 - 4 239 66
BC009007.1 - 7 - 8 807 3582639 66
Z37164.1 - 7 - 8 853 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600001986.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cct8

Panther Classification:

Molecular Function -

chaperonin

Gene Ontology Categories:

Function(s) Process(es)

protein folding
binding of sperm to zona pellucida
pore complex assembly
toxin transport
nucleotide binding
protein binding
ATP binding
unfolded protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600015436.mm.1 NM_009840.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600015436.mm.1 NM_009840.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600015436.mm.1 TC1600001986.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600015435.mm.1 NM_009840.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600015435.mm.1 NM_009840.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600015435.mm.1 TC1600001986.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline