This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01170924_m1
See other NEIL1 GE Assays ›
Gene Symbol:
Neil1
Gene Name:
nei endonuclease VIII-like 1 (E. coli)
Gene Aliases:
2810450N13Rik, Nei1
Chromosome Location:
Chr.9: 57142788 - 57148675 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01170924_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Neil1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK013322.1 - 1 - 2 47 66
AK155405.1 - 1 - 2 41 66
BC034873.1 - 1 - 2 221 66
BC043297.1 - 1 - 2 140 5320651 66

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK013322.1, AK155405.1, BC043297.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Panther Classification:

Molecular Function -

DNA glycosylase

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900019802.mm.1 NM_028347.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900019802.mm.1 NM_028347.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900019802.mm.1 TC0900002365.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900019803.mm.1 NM_028347.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900019803.mm.1 NM_028347.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900019803.mm.1 TC0900002365.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900019806.mm.1 TC0900002365.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 385524 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline