This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01170992_m1
See other BCO2 GE Assays ›
Gene Symbol:
Bco2
Gene Name:
beta-carotene oxygenase 2
Gene Aliases:
B-diox-II, Bcdo2, Bcmo2, CMO2
Chromosome Location:
Chr.9: 50533087 - 50555220 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01170992_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bco2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133217.3 NP_573480.1 6 - 7 881 98
GenBank mRNA AJ290392.1 - 5 - 6 724 98
BC107006.2 - 5 - 6 724 40046913 98
BC107007.2 - 5 - 6 724 40046914 98

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900002269.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Bco2

Panther Classification:

Molecular Function -

oxygenase

Gene Ontology Categories:

Function(s) Process(es)

carotenoid metabolic process
carotene metabolic process
carotene catabolic process
retinoic acid metabolic process
retinal metabolic process
regulation of mitochondrial membrane potential
oxidation-reduction process
regulation of reactive oxygen species metabolic process
oxidoreductase activity
oxidoreductase activity, acting on single donors with incorporation of molecular oxygen, incorporation of two atoms of oxygen
metal ion binding
dioxygenase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900018863.mm.1 NM_133217.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900018863.mm.1 NM_133217.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900018863.mm.1 TC0900002269.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-bjcbn:80/100.66.75.54:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0