This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01171137_g1
See other PIF1 GE Assays ›
Gene Symbol:
Pif1
Gene Name:
PIF1 5'-to-3' DNA helicase homolog (S. cerevisiae)
Gene Aliases:
4631410M14, AI449441
Chromosome Location:
Chr.9: 65587147 - 65595967 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01171137_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pif1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172453.3 NP_766041.1 11 - 12 2497 99
GenBank mRNA AK049353.1 - 11 - 12 2499 99
AK154072.1 - 11 - 12 2543 99
AY100322.1 - 12 - 13 2210 99
AY498715.1 - 11 - 12 2497 99
BC024481.1 - 5 - 6 631 99
BC046611.1 - 12 - 13 2135 6396273 99
BC060108.1 - 9 - 10 2708 99

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900000847.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pif1

Panther Classification:

Molecular Function -

DNA helicase

Gene Ontology Categories:

Function(s) Process(es)

telomere maintenance
DNA repair
DNA recombination
cellular response to DNA damage stimulus
regulation of telomere maintenance
DNA duplex unwinding
negative regulation of telomerase activity
nucleotide binding
magnesium ion binding
DNA binding
DNA helicase activity
helicase activity
ATP binding
hydrolase activity
single-stranded DNA-dependent ATP-dependent DNA helicase activity
ATP-dependent 5'-3' DNA/RNA helicase activity
telomeric DNA binding
ATP-dependent 5'-3' DNA helicase activity

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26588473 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26588472 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26588475 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P467448 BC060108.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P467448 AY498715.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P467448 AY100322.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P467448 AK154072.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P467448 AK049353.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P467448 BC060108.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P467448 AY498715.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P467448 AY100322.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P467448 AK154072.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P467448 AK049353.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 749362 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 382819 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 108904 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 252025 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 704226 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 135704 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152260 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P467448 BC060108.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P467448 AY498715.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P467448 AY100322.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P467448 AK154072.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P467448 AK049353.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P467448 BC060108.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P467448 AY498715.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P467448 AY100322.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P467448 AK154072.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P467448 AK049353.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26588472 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26588476 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26588480 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26588469 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26588486 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26588468 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26588475 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26588479 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26588474 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26588490 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26588482 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26588488 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26588478 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26588471 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26588484 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P152260 - -
Whole Mouse Genome G4122A A_51_P467448 BC060108.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P467448 AY498715.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P467448 AY100322.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P467448 AK154072.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P467448 AK049353.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P467448 BC060108.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P467448 AY498715.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P467448 AY100322.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P467448 AK154072.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P467448 AK049353.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133406 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P152260 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07710269 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07710281 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07710284 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26588475 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26588473 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26588472 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588543 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588507 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588510 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588531 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588486 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588522 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588455 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588432 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588526 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588524 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588551 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588467 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588547 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588449 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588482 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588517 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588503 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588559 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588508 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588474 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588504 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588492 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588513 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588439 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588466 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588494 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588490 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588488 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588429 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588535 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588459 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588545 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588472 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588434 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588495 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588511 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588468 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588447 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588480 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588476 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588457 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588452 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588471 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588479 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588454 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588501 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588475 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588453 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588534 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588431 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588518 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588435 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588433 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588533 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588484 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588442 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588546 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588554 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588436 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588469 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588516 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588514 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588444 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588438 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588548 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588448 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588445 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588426 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588464 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26588499 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588460 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07589514 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588524 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588486 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588488 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588464 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08093093 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588525 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588531 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588475 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588534 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07979574 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588461 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588468 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588518 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588463 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588466 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588476 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07990204 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07990201 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588480 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588533 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588503 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588496 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588513 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07710248 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588501 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588471 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588495 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588510 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08187818 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07476585 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07990202 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588469 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588508 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588491 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588499 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588482 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588506 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07979582 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588479 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588519 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07476584 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07899829 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588494 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588535 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588472 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588467 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588516 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588511 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588490 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26588514 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10586431 - -
Affymetrix Mouse Gene 1.0 ST Array 10586419 - -
Affymetrix Mouse Gene 1.0 ST Array 10586424 - -
Affymetrix Mouse Gene 1.0 ST Array 10586418 - -
Affymetrix Mouse Gene 1.0 ST Array 10586428 - -
Affymetrix Mouse Gene 1.0 ST Array 10586429 - -
Affymetrix Mouse Gene 1.0 ST Array 10586432 - -
Affymetrix Mouse Gene 1.0 ST Array 10586426 - -
Affymetrix Mouse Gene 1.0 ST Array 10586420 - -
Affymetrix Mouse Gene 1.0 ST Array 10586427 - -
Affymetrix Mouse Gene 1.0 ST Array 10586422 - -
Affymetrix Mouse Gene 1.0 ST Array 10586423 - -
Affymetrix Mouse Gene 1.0 ST Array 10586430 - -
Affymetrix Mouse Gene 1.0 ST Array 10586417 - -
Affymetrix Mouse Gene 1.0 ST Array 10586425 - -
Affymetrix Mouse Gene 1.0 ST Array 10586421 - -
Affymetrix HT MG-430 PM Array Plate 1439297_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1438149_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1436738_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10586416 - -
Mouse Genome 430 2.0 Array 1439297_PM_at - -
Mouse Genome 430 2.0 Array 1439297_at - -
Mouse Genome 430 2.0 Array 1436738_PM_at - -
Mouse Genome 430 2.0 Array 1438149_at - -
Mouse Genome 430 2.0 Array 1436738_at - -
Mouse Genome 430 2.0 Array 1438149_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 208084_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 208084_PM_at - -
Affymetrix Mouse Clariom S Array PSR0900006494.mm.1 NM_172453.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900006494.mm.1 NM_172453.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900006494.mm.1 TC0900000847.mm.1 MTA TC
Mouse Expression Set 430 1436738_PM_at - -
Mouse Expression Set 430 1436738_at - -
Mouse Expression Set 430 1439297_at - -
Mouse Expression Set 430 1439297_PM_at - -
Mouse Expression Set 430 1438149_PM_at - -
Mouse Expression Set 430 1438149_at - -
Murine Genome U74v2 Set 167721_PM_at - -
Murine Genome U74v2 Set 167721_at - -
Affymetrix HT Mouse Genome 430B Array 1438149_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1438149_at - -
Affymetrix HT Mouse Genome 430B Array 1436738_at - -
Affymetrix HT Mouse Genome 430B Array 1439297_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1439297_at - -
Affymetrix HT Mouse Genome 430B Array 1436738_PM_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 104670079 - -
MouseWG-6 v2 Expression BeadChip 4850451 - -
Illumina mouse-6 v1.0 expression beadchip 4850451 - -
Illumina mouse-6 v1.0 expression beadchip 104670079 - -
Illumina mouseRef-8 v1.0 expression beadchip 0004850451 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1258710 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1249239 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1258710 - -
mouse-6 v1.1 (Illumina) scl36803.7_641-S - -
Illumina Sentrix mouse-6.v1 bead array 4850451 - -
Illumina Sentrix mouse-6.v1 bead array 104670079 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl36803.7_641-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 104670079 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 4850451 - -
MouseRef-8 v2 Expression BeadChip ILMN_2714678 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1249239 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2714678 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0