This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01171139_g1
See other PIF1 GE Assays ›
Gene Symbol:
Pif1
Gene Name:
PIF1 5'-to-3' DNA helicase homolog (S. cerevisiae)
Gene Aliases:
4631410M14, AI449441
Chromosome Location:
Chr.9: 65587147 - 65595967 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01171139_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pif1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172453.3 NP_766041.1 3 - 4 1451 90
GenBank mRNA AK049353.1 - 3 - 4 1453 90
AK154072.1 - 3 - 4 1464 90
AY100322.1 - 4 - 5 1164 90
AY498715.1 - 3 - 4 1451 90
BC046611.1 - 4 - 5 1089 6396273 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900000847.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pif1

Panther Classification:

Molecular Function -

DNA helicase

Gene Ontology Categories:

Function(s) Process(es)

telomere maintenance
DNA repair
DNA recombination
cellular response to DNA damage stimulus
regulation of telomere maintenance
DNA duplex unwinding
negative regulation of telomerase activity
nucleotide binding
magnesium ion binding
DNA binding
DNA helicase activity
helicase activity
ATP binding
hydrolase activity
single-stranded DNA-dependent ATP-dependent DNA helicase activity
ATP-dependent 5'-3' DNA/RNA helicase activity
telomeric DNA binding
ATP-dependent 5'-3' DNA helicase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900006480.mm.1 NM_172453.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900006480.mm.1 NM_172453.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900006480.mm.1 TC0900000847.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900006485.mm.1 NM_172453.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900006485.mm.1 NM_172453.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900006485.mm.1 TC0900000847.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900006481.mm.1 NM_172453.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900006481.mm.1 NM_172453.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900006481.mm.1 TC0900000847.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900006479.mm.1 NM_172453.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900006479.mm.1 NM_172453.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900006479.mm.1 TC0900000847.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900006477.mm.1 NM_172453.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900006477.mm.1 NM_172453.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900006477.mm.1 TC0900000847.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0