This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01171182_m1
See other COMT GE Assays ›
Gene Symbol:
Comt
Gene Name:
catechol-O-methyltransferase
Gene Aliases:
Comt1, D16Wsu103e, D330014B15Rik
Chromosome Location:
Chr.16: 18406882 - 18426716 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01171182_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Comt
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001111062.1 NP_001104532.1 1 - 2 360 144

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001111062.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

catecholamine metabolic process
female pregnancy
learning
short-term memory
estrogen metabolic process
catechol-containing compound metabolic process
cellular response to phosphate starvation
methylation
negative regulation of renal sodium excretion
neurotransmitter catabolic process
dopamine metabolic process
dopamine catabolic process
negative regulation of dopamine metabolic process
response to pain
multicellular organismal reproductive process
negative regulation of smooth muscle cell proliferation
positive regulation of homocysteine metabolic process
regulation of sensory perception of pain
magnesium ion binding
methyltransferase activity
O-methyltransferase activity
catechol O-methyltransferase activity
transferase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600010854.mm.1 NM_001111062.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600010854.mm.1 NM_001111062.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600010854.mm.1 NM_007744.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600010854.mm.1 NM_007744.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600010854.mm.1 TC1600001316.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600010853.mm.1 NM_001111062.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600010853.mm.1 NM_001111062.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600010853.mm.1 NM_007744.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600010853.mm.1 NM_007744.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600010853.mm.1 TC1600001316.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600010852.mm.1 NM_001111062.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600010852.mm.1 NM_001111062.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600010852.mm.1 NM_007744.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600010852.mm.1 NM_007744.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600010852.mm.1 TC1600001316.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600010851.mm.1 NM_001111062.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600010851.mm.1 NM_001111062.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600010851.mm.1 TC1600001316.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0