This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01171276_mH
See other PLD3 GE Assays ›
Gene Symbol:
Pld3
Gene Name:
phospholipase D family, member 3
Gene Aliases:
Sam-9
Chromosome Location:
Chr.7: 27531645 - 27553213 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01171276_mH
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pld3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011116.1 NP_035246.1 9 - 10 1085 76
GenBank mRNA AF026124.1 - 9 - 10 1085 76
AK153508.1 - 7 - 8 774 76
BC068170.1 - 9 - 10 1081 76
BC076586.1 - 9 - 10 1149 30604186 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700002669.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pld3

Panther Classification:

Molecular Function -

phospholipase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
lipid catabolic process
catalytic activity
phospholipase D activity
hydrolase activity
N-acylphosphatidylethanolamine-specific phospholipase D activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700023299.mm.1 NM_011116.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700023299.mm.1 TC0700002669.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline