This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01172918_g1
See other CREB3L3 GE Assays ›
Gene Symbol:
Creb3l3
Gene Name:
cAMP responsive element binding protein 3-like 3
Gene Aliases:
BC010786, CREB-H, D10Bur1e
Chromosome Location:
Chr.10: 81084324 - 81098872 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01172918_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Creb3l3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145365.3 NP_663340.1 4 - 5 575 87
GenBank mRNA AF392874.1 - 2 - 3 226 87
AK050186.1 - 2 - 3 217 87
AK077258.1 - 4 - 5 576 87
BC010786.1 - 2 - 3 270 4211480 87
BC028820.1 - 2 - 3 247 5096773 87

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000002525.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Creb3l3

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
response to unfolded protein
endoplasmic reticulum unfolded protein response
positive regulation of transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
transcription regulatory region sequence-specific DNA binding
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
cAMP response element binding
protein homodimerization activity
sequence-specific DNA binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000018885.mm.1 NM_145365.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000018885.mm.1 NM_145365.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000018885.mm.1 TC1000002525.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000018876.mm.1 NM_145365.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000018876.mm.1 NM_145365.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000018876.mm.1 TC1000002525.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000018879.mm.1 NM_145365.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000018879.mm.1 NM_145365.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000018879.mm.1 TC1000002525.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000018886.mm.1 NM_145365.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000018886.mm.1 NM_145365.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000018886.mm.1 TC1000002525.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000018878.mm.1 NM_145365.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000018878.mm.1 NM_145365.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000018878.mm.1 TC1000002525.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000018883.mm.1 NM_145365.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000018883.mm.1 NM_145365.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000018883.mm.1 TC1000002525.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000018875.mm.1 NM_145365.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000018875.mm.1 NM_145365.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000018875.mm.1 TC1000002525.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-6qxl5:80/100.66.77.227:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline