This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01172976_g1
See other HCST GE Assays ›
Gene Symbol:
Hcst
Gene Name:
hematopoietic cell signal transducer
Gene Aliases:
DAP10, KAP10
Chromosome Location:
Chr.7: 30417712 - 30419854 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01172976_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hcst
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011827.3 NP_035957.2 2 - 3 171 62
GenBank mRNA AF072846.1 - 2 - 3 63 62
AF172930.1 - 2 - 3 171 62
AK008005.1 - 2 - 3 170 62
BC145545.1 - 2 - 3 162 9054109 62
BC145824.1 - 2 - 3 162 40131127 62
BC145826.1 - 2 - 3 162 40131129 62

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hcst

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
positive regulation of phosphatidylinositol 3-kinase signaling
receptor binding
protein binding
phosphatidylinositol 3-kinase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0