This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01172986_m1
See other YIF1B GE Assays ›
Gene Symbol:
Yif1b
Gene Name:
Yip1 interacting factor homolog B (S. cerevisiae)
Gene Aliases:
9430029K10Rik, Yip1b
Chromosome Location:
Chr.7: 29238308 - 29247532 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01172986_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Yif1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029887.3 NP_084163.2 1 - 2 120 91
GenBank mRNA AK020444.1 - 1 - 2 91 91

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700000526.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Yif1b

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700004476.mm.1 TC0700000526.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700004476.mm.1 NM_001110201.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700004476.mm.1 NM_001110201.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700004476.mm.1 NM_029887.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700004476.mm.1 NM_029887.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-r7bw6:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0