This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01173295_m1
See other PRMT2 GE Assays ›
Gene Symbol:
Prmt2
Gene Name:
protein arginine N-methyltransferase 2
Gene Aliases:
AI504737, Hrmt1l1
Chromosome Location:
Chr.10: 76207226 - 76237865 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01173295_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prmt2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK077456.1 - 1 - 2 84 102
AK140200.1 - 1 - 2 111 102
AK145829.1 - 1 - 2 84 102
AK146754.1 - 1 - 2 83 102
AK146966.1 - 1 - 2 91 102
AK159186.1 - 1 - 2 90 102
BY727303.1 - 1 - 2 110 102

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK140200.1, AK145829.1, AK146966.1, AK159186.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Mus musculus 1700 Probe spans exons

Panther Classification:

Molecular Function -

protein modifying enzyme

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 622698 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0