This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01173297_m1
See other PRMT2 GE Assays ›
Gene Symbol:
Prmt2
Gene Name:
protein arginine N-methyltransferase 2
Gene Aliases:
AI504737, Hrmt1l1
Chromosome Location:
Chr.10: 76207226 - 76237865 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01173297_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prmt2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AF169620.1 - 6 - 7 775 69
AK077456.1 - 6 - 7 832 69
AK144412.1 - 6 - 7 830 69
AK145829.1 - 6 - 7 832 69
AK146966.1 - 6 - 7 839 69
AK159186.1 - 6 - 7 838 69
BC088997.1 - 6 - 7 789 69
BC122563.1 - 5 - 6 763 40107557 69
BC125275.1 - 5 - 6 763 40107558 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000002398.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

protein modifying enzyme

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000017197.mm.1 NM_001077638.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000017197.mm.1 NM_133182.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000017197.mm.1 TC1000002398.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-r7bw6:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0