This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01173300_m1
See other PRMT2 GE Assays ›
Gene Symbol:
Prmt2
Gene Name:
protein arginine N-methyltransferase 2
Gene Aliases:
AI504737, Hrmt1l1
Chromosome Location:
Chr.10: 76207226 - 76237865 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01173300_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prmt2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK077456.1 - 9 - 10 1267 105
AK140200.1 - 10 - 11 1355 105
AK144412.1 - 9 - 10 1265 105
AK145829.1 - 9 - 10 1267 105
AK146754.1 - 10 - 11 1328 105
AK159186.1 - 9 - 10 1273 105
BC088997.1 - 9 - 10 1224 105
BC122563.1 - 8 - 9 1198 40107557 105
BC125275.1 - 8 - 9 1198 40107558 105

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000002398.mm.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

protein modifying enzyme

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000017190.mm.1 NM_001077638.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000017190.mm.1 NM_133182.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000017190.mm.1 TC1000002398.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0