This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01173827_m1
See other GTF2A2 GE Assays ›
Gene Symbol:
Gtf2a2
Gene Name:
general transcription factor II A, 2
Gene Aliases:
TFIIA-gamma, TfIIg, Tfiia2
Chromosome Location:
Chr.9: 70011851 - 70022866 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01173827_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gtf2a2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001282083.1 NP_001269012.1 1 - 2 409 151
GenBank mRNA AK050621.1 - 1 - 2 387 151

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK050621.1, NM_001282083.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
positive regulation of sequence-specific DNA binding transcription factor activity
RNA polymerase II transcriptional preinitiation complex assembly
RNA polymerase II repressing transcription factor binding
transcription coactivator activity
transcription factor binding
TBP-class protein binding
protein homodimerization activity
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900007142.mm.1 NM_001282083.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900007142.mm.1 TC0900000923.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900007145.mm.1 NM_001039519.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900007145.mm.1 NM_001282083.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900007145.mm.1 NM_001282084.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900007145.mm.1 NR_104093.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900007145.mm.1 TC0900000923.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline